2-Fluoro-5-(hydroxymethyl)-6-picoline 97%

Supplier: Ambeed
Warning

Synonyms: (6-Fluoro-2-methylpyridin-3-yl)methanol, 6-Fluoro-3-hydroxymethyl-2-methylpyridine

A159424-100MG A159424-1G A159424-250MG A159424-5G
77110-582EA 41.38 USD
77110-582 77110-584 77110-586 77110-588
2-Fluoro-5-(hydroxymethyl)-6-picoline 97%
2-Fluoro-5-(hydroxymethyl)-6-picoline

Precaución: for Research Use Only

Formula: C₇H₈FNO
MW: 141.1400 g/mol
MDL Number: MFCD05662401
CAS Number: 884494-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR