5-Methyl-1,3-cyclohexanedione 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methylcyclohexane-1,3-dione

A159241-1G A159241-5G
77110-580EA 7.43 USD
77110-580 77611-532
5-Methyl-1,3-cyclohexanedione 98%
5-Methyl-1,3-cyclohexanedione

Precaución: for Research Use Only

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
MDL Number: MFCD00010379
CAS Number: 4341-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR