4-Benzyloxy-2,3-difluorophenylboronic acid 97%

Supplier: Ambeed
Warning

A159235-1G A159235-250MG A159235-5G
77110-574EA 25.75 USD
77110-574 77110-576 77110-578
4-Benzyloxy-2,3-difluorophenylboronic acid 97%
4-Benzyloxy-2,3-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₂O₃
MDL Number: MFCD07644469
CAS Number: 156635-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR