2-Bromonaphthalene 98%

Supplier: Ambeed
Warning

A157955-1000G A157955-100G A157955-10G A157955-1G A157955-500G A157955-5G A157955-25G
77110-550EA 408.91 USD
77110-550 77110-552 77110-554 77110-556 77110-558 77110-560 77319-108
2-Bromonaphthalene 98%
2-Bromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₇Br
MW: 207.07 g/mol
MDL Number: MFCD00004051
CAS Number: 580-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR