Sunitinib 98%

Supplier: Ambeed
Danger

Synonyms: (Z)-N-(2-(Diethylamino)ethyl)-5-((5-fluoro-2-oxoindolin-3-ylidene)methyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide

A157426-1G A157426-1MG A157426-250MG A157426-5G A157426-100MG
77110-538EA 39.26 USD
77110-538 77110-540 77110-542 77110-544 77615-282
Sunitinib 98%
Sunitinib

Precaución: for Research Use Only

Formula: C₂₂H₂₇FN₄O₂
MW: 398.47 g/mol
MDL Number: MFCD09260778
CAS Number: 557795-19-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR