3-Amino-4-fluorophenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Borono-2-fluoroaniline, Boronic acid, B-(3-amino-4-fluorophenyl)-, 3-Amino-4-fluorobenzeneboronic acid

A157302-1G A157302-250MG A157302-100MG
77110-534EA 51.51 USD
77110-534 77110-536 77685-296
3-Amino-4-fluorophenylboronic acid 95%
3-Amino-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BFNO₂
MDL Number: MFCD03092925
CAS Number: 873566-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR