2-Hydroxy-3-nitro-6-picoline 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Methyl-3-nitro-2(1H)-pyridinone, 2-Hydroxy-6-methyl-3-nitropyridine, 6-Hydroxy-5-nitro-2-picoline, 6-Methyl-3-nitropyridin-2-ol, 6-Methyl-3-nitro-2-pyridone, 2-Methyl-5-nitro-6-pyridone, 6-Hydroxy-2-methyl-5-nitropyridine
A156001-100G
A156001-10G
A156001-1G
A156001-250MG
A156001-25G
A156001-5G
77110-480EA
270.88
USD
77110-480
77110-482
77110-484
77110-486
77110-488
77110-490
2-Hydroxy-3-nitro-6-picoline 95%
2-Hydroxy-3-nitro-6-picoline
Precaución: for Research Use Only
Formula:
C₆H₆N₂O₃ MW: 154.13 |
MDL Number:
MFCD03001414 CAS Number: 39745-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...