Ethyl-2-chloro-6-benzothiazolecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-chloro-6-benzothiazolecarboxylate

A155247-10G A155247-1G A155247-25G A155247-5G
77110-460EA 264.17 USD
77110-460 77110-462 77110-464 77110-466
Ethyl-2-chloro-6-benzothiazolecarboxylate 97%
Ethyl-2-chloro-6-benzothiazolecarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO₂S
MW: 241.6900 g/mol
MDL Number: MFCD08443995
CAS Number: 78485-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR