9-Allyl-9H-carbazole 95%

Supplier: Ambeed
Warning

A154916-250MG A154916-1G
77110-458EA 15.92 USD
77110-458 77685-280
9-Allyl-9H-carbazole 95%
9-Allyl-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₅H₁₃N
MW: 207.27 g/mol
MDL Number: MFCD00093414
CAS Number: 3998-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR