4-Amino-2-methylpyridine 97%

Supplier: Ambeed
Warning

A154495-100G A154495-10G A154495-1G A154495-25G A154495-5G
77110-446EA 136.99 USD
77110-446 77110-448 77110-450 77110-452 77110-454
4-Amino-2-methylpyridine 97%
4-Amino-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₈N₂
MDL Number: MFCD00186509
CAS Number: 18437-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR