25-Hydroxyvitamin D3 98%

Supplier: Ambeed
Danger

Synonyms: Calcifediol, (R,Z)-3-(2-((1R,3aS,7aR,E)-1-((R)-6-hydroxy-6-methylheptan-2-yl)-7a-methyloctahydro-4H-inden-4-ylidene)ethylidene)-4-methylenecyclohexan-1-ol

A154275-10MG A154275-25MG A154275-50MG A154275-5MG
77110-434EA 159.14 USD
77110-434 77110-436 77110-438 77110-440
25-Hydroxyvitamin D3 98%
25-Hydroxyvitamin D3

Precaución: for Research Use Only

Formula: C₂₇H₄₄O₂
MW: 400.6 g/mol
MDL Number: MFCD00867077
CAS Number: 19356-17-3
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR