2-Mercapto-5-methyl-1,3,4-thiadiazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-thio-1,3,4-thiadiazole, 5-Methyl-1,3,4-thiadiazole-2-thiol

A153687-1000G A153687-100G A153687-10G A153687-25G A153687-500G
77110-404EA 108.22 USD
77110-404 77110-406 77110-408 77110-410 77110-412
2-Mercapto-5-methyl-1,3,4-thiadiazole 97%
2-Mercapto-5-methyl-1,3,4-thiadiazole

Precaución: for Research Use Only

Formula: C₃H₄N₂S₂
MW: 132.21 g/mol
MDL Number: MFCD00038349
CAS Number: 29490-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR