(S)-3-(Allylthio)-2-aminopropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: H-D-S-Allyl-Cys-OH

A152522-10G A152522-1G A152522-250MG A152522-25G A152522-5G
77110-372EA 231.28 USD
77110-372 77110-374 77110-376 77110-378 77110-380
(S)-3-(Allylthio)-2-aminopropanoic acid 95%
(S)-3-(Allylthio)-2-aminopropanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₁NO₂S
MW: 161.22 g/mol
MDL Number: MFCD00210474
CAS Number: 770742-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR