2-Amino-5-nitro-6-picoline 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2-methyl-3-nitropyridine, 2-Amino-5-nitro-6-methylpyridine, 2-Amino-6-methyl-5-nitropyridine, 6-Amino-3-nitro-2-picoline, 6-Methyl-5-nitro-2-pyridylamine

A152464-1G A152464-25G A152464-5G
77110-366EA 5.15 USD
77110-366 77110-368 77110-370
2-Amino-5-nitro-6-picoline 97%
2-Amino-5-nitro-6-picoline

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.14
MDL Number: MFCD00053582
CAS Number: 22280-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR