1-(2-Amino-4-methoxy-3-methylphenyl)ethanone 97%
Supplier: Ambeed
|
Warning
|
A152219-1G
A152219-250MG
A152219-5G
77110-360EA
28.84
USD
77110-360
77110-362
77110-364
1-(2-Amino-4-methoxy-3-methylphenyl)ethanone 97%
1-(2-Amino-4-methoxy-3-methylphenyl)ethanone
Precaución: for Research Use Only
Formula:
C₁₀H₁₃NO₂ |
MDL Number:
MFCD11042290 CAS Number: 912347-94-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...