(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dapoxetine HCl, (S)-N,N-Dimethyl-alpha-[2-(1-naphthyloxy)ethyl]benzylamine hydrochloride, Dapoxetine hydrochloride, (S)-N,N-Dimethyl-3-(1-naphthyloxy)-1-phenylpropylamine hydrochloride
A151531-100G
A151531-100MG
A151531-1G
A151531-50MG
A151531-5G
A151531-25G
77110-324EA
356.38
USD
77110-324
77110-326
77110-328
77110-330
77110-332
77685-259
(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride 97%
(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₂₁H₂₄ClNO |
MDL Number:
MFCD08272809 CAS Number: 129938-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...