2,2-Dimethylsuccinic anhydride 98%

Supplier: Ambeed
Warning

Synonyms: 2,2-Dimethylsuccinicanhydride

A151012-100G A151012-10G A151012-1G A151012-25G A151012-5G
77110-302EA 122.01 USD
77110-302 77110-304 77110-306 77110-308 77110-310
2,2-Dimethylsuccinic anhydride 98%
2,2-Dimethylsuccinic anhydride

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.1259 g/mol
MDL Number: MFCD00022608
CAS Number: 17347-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR