Fmoc-N-methyl-D-norleucine 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)hexanoic acid

A150969-1G A150969-250MG
77110-298EA 101.85 USD
77110-298 77110-300
Fmoc-N-methyl-D-norleucine 95%
Fmoc-N-methyl-D-norleucine

Precaución: for Research Use Only

Formula: C₂₂H₂₅NO₄
MW: 367.438 g/mol
MDL Number: MFCD07366865
CAS Number: 1217482-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR