DL-Methionine 98%

Supplier: Ambeed
Warning

A150589-1000G A150589-100G A150589-25G A150589-500G
77110-278EA 29.87 USD
77110-278 77110-280 77110-282 77110-284
DL-Methionine 98%
DL-Methionine

Precaución: for Research Use Only

Formula: C₅H₁₁NO₂S
MDL Number: MFCD00063096
CAS Number: 59-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR