1-Cyclopropylpiperazine dihydrochloride 95%

Supplier: Ambeed
Warning

A150354-1G A150354-250MG
77110-264EA 92.7 USD
77110-264 77110-266
1-Cyclopropylpiperazine dihydrochloride 95%
1-Cyclopropylpiperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄N₂·2HCl
MW: 199.12 g/mol
MDL Number: MFCD00040810
CAS Number: 139256-79-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR