2,4-Dibromo-5-fluoro-3-nitropyridine 95%

Supplier: Ambeed
Warning

A150228-100MG A150228-250MG A150228-1G
77110-252EA 81.69 USD
77110-252 77110-254 77611-392
2,4-Dibromo-5-fluoro-3-nitropyridine 95%
2,4-Dibromo-5-fluoro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃BrN₂O₂
MW: 299.88 g/mol
MDL Number: MFCD05662393
CAS Number: 884494-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR