N1,N6-Bis(3-phenylallylidene)hexane-1,6-diamine 95%

Supplier: Ambeed
Warning

A149910-10G A149910-5G
77110-234EA 8.49 USD
77110-234 77110-236
N1,N6-Bis(3-phenylallylidene)hexane-1,6-diamine 95%
N1,N6-Bis(3-phenylallylidene)hexane-1,6-diamine

Precaución: for Research Use Only

Formula: C₂₄H₂₈N₂
MW: 344.49 g/mol
MDL Number: MFCD00046784
CAS Number: 140-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR