1-(4-Bromo-5-fluoro-2-hydroxyphenyl)ethanone 95%

Supplier: Ambeed
Warning

A149891-100MG A149891-1G A149891-250MG
77110-228EA 14.42 USD
77110-228 77110-230 77110-232
1-(4-Bromo-5-fluoro-2-hydroxyphenyl)ethanone 95%
1-(4-Bromo-5-fluoro-2-hydroxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MDL Number: MFCD11847403
CAS Number: 865449-63-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR