2-Chloro-4,6-dimethylpyrimidine 97%

Supplier: Ambeed
Warning

A149139-100G A149139-1G A149139-25G A149139-500G A149139-5G
77110-214EA 132.62 USD
77110-214 77110-216 77110-218 77110-220 77110-222
2-Chloro-4,6-dimethylpyrimidine 97%
2-Chloro-4,6-dimethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
MDL Number: MFCD00023199
CAS Number: 4472-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR