(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid 98%

Supplier: Ambeed
Warning

A149022-10G A149022-1G A149022-250MG A149022-5G A149022-25G
77110-206EA 219.61 USD
77110-206 77110-208 77110-210 77110-212 77685-246
(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid 98%
(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyric acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.21 g/mol
MDL Number: MFCD00070368
CAS Number: 59554-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR