H-Arg(MTR)-OH 97%

Supplier: Ambeed
Warning

A148764-10G A148764-1G A148764-5G
77110-180EA 22.66 USD
77110-180 77110-182 77110-184
H-Arg(MTR)-OH 97%
H-Arg(MTR)-OH

Precaución: for Research Use Only

Formula: C₁₆H₂₆N₄O₅S
MDL Number: MFCD00057846
CAS Number: 80745-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR