(S)-3-Amino-3-(3-nitrophenyl)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: (S)-3-Amino-3-(3-nitrophenyl)propanoic acid

A148728-1G A148728-250MG
77110-176EA 109.28 USD
77110-176 77110-178
(S)-3-Amino-3-(3-nitrophenyl)propionic acid 97%
(S)-3-Amino-3-(3-nitrophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
MDL Number: MFCD04113693
CAS Number: 734529-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR