4-Fluoroaniline hydrochloride 98+%

Supplier: Ambeed
Warning

A148499-1G A148499-5G
77110-156EA 10.31 USD
77110-156 77110-158
4-Fluoroaniline hydrochloride 98+%
4-Fluoroaniline hydrochloride

Precaución: for Research Use Only

Formula: C₆H₆FN·HCl
MDL Number: MFCD00035500
CAS Number: 2146-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR