2-((Diphenylmethylene)amino)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: N-(Diphenylmethylene)aminoacetonitrile

A148119-100G A148119-1G A148119-25G A148119-500G A148119-5G
77110-134EA 124.13 USD
77110-134 77110-136 77110-138 77110-140 77110-142
2-((Diphenylmethylene)amino)acetonitrile 97%
2-((Diphenylmethylene)amino)acetonitrile

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂
MW: 220.2700 g/mol
MDL Number: MFCD00009970
CAS Number: 70591-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR