1-Bromo-4-chloroisoquinoline 97%

Supplier: Ambeed
Warning

A148016-1G A148016-250MG A148016-5G
77110-124EA 106.08 USD
77110-124 77110-126 77110-128
1-Bromo-4-chloroisoquinoline 97%
1-Bromo-4-chloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MDL Number: MFCD08437045
CAS Number: 925890-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR