5-Bromo-1H-indene 95%

Supplier: Ambeed
Warning

A147747-100MG A147747-1G A147747-250MG
77110-108EA 41.38 USD
77110-108 77110-110 77110-112
5-Bromo-1H-indene 95%
5-Bromo-1H-indene

Precaución: for Research Use Only

Formula: C₉H₇Br
MW: 195.056 g/mol
MDL Number: MFCD03659729
CAS Number: 75476-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR