Methyl-2-amino-4,5-difluorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-amino-4,5-difluorobenzoate

A147427-100G A147427-10G A147427-1G A147427-25G A147427-5G
77110-096EA 138.98 USD
77110-096 77110-098 77110-100 77110-102 77110-104
Methyl-2-amino-4,5-difluorobenzoate 98%
Methyl-2-amino-4,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MW: 187.15 g/mol
MDL Number: MFCD09264511
CAS Number: 207346-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR