9H-Fluoren-9-ylmethyl-4-aminopiperidine-1-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (9H-Fluoren-9-yl)methyl 4-aminopiperidine-1-carboxylate hydrochloride

A146056-25G A146056-5G A146056-1G
77110-052EA 665.37 USD
77110-052 77110-054 77566-976
9H-Fluoren-9-ylmethyl-4-aminopiperidine-1-carboxylate hydrochloride 98%
9H-Fluoren-9-ylmethyl-4-aminopiperidine-1-carboxylate hydrochloride

Precaución: For research use only.

Formula: C₂₀H₂₃ClN₂O₂
MW: 358.87 g/mol
MDL Number: MFCD03001675
CAS Number: 811841-89-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR