(S)-1-Carbobenzoxy-3-pyrrolidinol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-1-Cbz-3-pyrrolidinol, (S)-1-Carbobenzoxy-3-hydroxypyrrolidine, (S)-(+)-1-Benzyloxycarbonyl-3-hydroxypyrrolidine, Benzyl (S)-3-hydroxypyrrolidine-1-carboxylate

A145605-100G A145605-10G A145605-1G A145605-250MG A145605-25G A145605-500G A145605-5G
77110-024EA 164.8 USD
77110-024 77110-026 77110-028 77110-030 77110-032 77110-034 77110-036
(S)-1-Carbobenzoxy-3-pyrrolidinol 97%
(S)-1-Carbobenzoxy-3-pyrrolidinol

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD07784363
CAS Number: 100858-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR