2-Methoxy-3-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxypyridine-3-boronic acid

A145477-10G A145477-1G A145477-250MG A145477-25G A145477-5G A145477-100G A145477-500G
77110-014EA 24.73 USD
77110-014 77110-016 77110-018 77110-020 77110-022 77611-336 77685-233
2-Methoxy-3-pyridineboronic acid 98%
2-Methoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₈BNO₃
MDL Number: MFCD03411572
CAS Number: 163105-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR