2-Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylic acid 98%

Supplier: Ambeed
Warning

A144783-1G A144783-250MG A144783-5G A144783-100MG
77109-980EA 124.13 USD
77109-980 77109-982 77109-984 77685-228
2-Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylic acid 98%
2-Methyl-4H-thieno[3,2-b]pyrrole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₂S
MW: 181.21 g/mol
MDL Number: MFCD08277221
CAS Number: 332099-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR