4-Bromo-8-fluoroquinoline 98%

Supplier: Ambeed
Warning

A144767-100MG A144767-1G A144767-250MG A144767-5G
77109-972EA 36.08 USD
77109-972 77109-974 77109-976 77109-978
4-Bromo-8-fluoroquinoline 98%
4-Bromo-8-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MW: 226.0451 g/mol
MDL Number: MFCD07700231
CAS Number: 927800-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR