Boc-Val-OMe 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-valine methyl ester, Boc-Val-OMe, N-(tert-Butoxycarbonyl)-L-valine methyll ester, (S)-Methyl 2-[(tert-Butoxycarbonyl)amino]-3-methylbutanoate, (S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-methylbutanoate, N-Boc-L-valine methyll ester

A144669-10G A144669-1G A144669-25G A144669-5G
77109-964EA 9.27 USD
77109-964 77109-966 77109-968 77109-970
Boc-Val-OMe 97%
Boc-Val-OMe

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00672510
CAS Number: 58561-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR