2,6-Difluoro-3-nitrobenzonitrile 98%

Supplier: Ambeed
Danger

A144579-1G A144579-25G A144579-5G A144579-100G
77109-958EA 6.37 USD
77109-958 77109-960 77109-962 77685-227
2,6-Difluoro-3-nitrobenzonitrile 98%
2,6-Difluoro-3-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂F₂N₂O₂
MDL Number: MFCD00042254
CAS Number: 143879-77-0
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR