L(+)-Tartaric acid 98%

Supplier: Ambeed
Warning

A144400-1000G A144400-100G A144400-500G
77109-944EA 15.92 USD
77109-944 77109-946 77109-948
L(+)-Tartaric acid 98%
L(+)-Tartaric acid

Precaución: for Research Use Only

Formula: C₄H₆O₆
MDL Number: MFCD00064207
CAS Number: 87-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR