Ciclopirox olamine 99%

Supplier: Ambeed
Warning

Synonyms: 6-Cyclohexyl-1-hydroxy-4-methylpyridin-2(1H)-one 2-aminoethanol salt

A144214-100MG A144214-1MG A144214-50MG A144214-5G A144214-1G
77109-932EA 8.49 USD
77109-932 77109-934 77109-936 77109-938 77615-152
Ciclopirox olamine 99%
Ciclopirox olamine

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₂·C₂H₇NO
MW: 268.3520 g/mol
MDL Number: MFCD00078997
CAS Number: 41621-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR