Ethyl-4,6-dihydroxynicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4,6-dihydroxynicotinate

A143933-10G A143933-1G A143933-25G A143933-5G
77109-924EA 85.94 USD
77109-924 77109-926 77109-928 77109-930
Ethyl-4,6-dihydroxynicotinate 98%
Ethyl-4,6-dihydroxynicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₄
MW: 183.17 g/mol
MDL Number: MFCD00834976
CAS Number: 6975-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR