4-Bromo-1,3-dimethoxybenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,4-dimethoxybenzene

A143868-100G A143868-10G A143868-5G A143868-25G A143868-500G
77109-906EA 48.41 USD
77109-906 77109-908 77109-910 77316-640 77615-150
4-Bromo-1,3-dimethoxybenzene 98%
4-Bromo-1,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Boiling Pt: 282
MDL Number: MFCD00009844
CAS Number: 17715-69-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR