Methyl 6-bromo-5-methylpicolinate 97%

Supplier: Ambeed
Warning

A143556-100MG A143556-1G A143556-250MG
77109-894EA 130.5 USD
77109-894 77109-896 77109-898
Methyl 6-bromo-5-methylpicolinate 97%
Methyl 6-bromo-5-methylpicolinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
MDL Number: MFCD18382716
CAS Number: 1209093-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR