2-Acetylnaphthalene 99%

Supplier: Ambeed
Warning

A143056-1000G A143056-100G A143056-500G A143056-25G
77109-886EA 88.58 USD
77109-886 77109-888 77109-890 77299-324
2-Acetylnaphthalene 99%
2-Acetylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₀O
MDL Number: MFCD00004108
CAS Number: 93-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR