DL-Lysine monohydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: (±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride

A142323-100G A142323-10G A142323-500G A142323-5G A142323-25G
77109-864EA 16.98 USD
77109-864 77109-866 77109-868 77109-870 77316-358
DL-Lysine monohydrochloride 95%
DL-Lysine monohydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₅ClN₂O₂
MW: 182.65 g/mol
MDL Number: MFCD00064563
CAS Number: 70-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR