2,5-Dimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A142275-100G A142275-10G A142275-500G A142275-5G A142275-25G
77109-856EA 38.12 USD
77109-856 77109-858 77109-860 77109-862 77316-356
2,5-Dimethoxybenzaldehyde 98%
2,5-Dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00003314
CAS Number: 93-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR