2-Nitroresorcinol 99%

Supplier: Ambeed
Warning

Synonyms: 2-Nitrobenzene-1,3-diol

A141918-25G A141918-5G
77109-840EA 33.95 USD
77109-840 77109-842
2-Nitroresorcinol 99%
2-Nitroresorcinol

Precaución: for Research Use Only

Formula: C₆H₅NO₄
MW: 155.11 g/mol
MDL Number: MFCD00007124
CAS Number: 601-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR