Nadifloxacin 98%

Supplier: Ambeed
Danger

Synonyms: 9-Fluoro-8-(4-hydroxypiperidin-1-yl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic acid, 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid

A140688-1G A140688-1MG A140688-250MG A140688-50MG A140688-5G
77109-796EA 66.84 USD
77109-796 77109-798 77109-800 77109-802 77109-804
Nadifloxacin 98%
Nadifloxacin

Precaución: for Research Use Only

Formula: C₁₉H₂₁FN₂O₄
MW: 360.39 g/mol
MDL Number: MFCD00865081
CAS Number: 124858-35-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR