2-Chloro-3,6-difluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A140399-100G
A140399-25G
A140399-5G
A140399-1G
77109-778EA
598.35
USD
77109-778
77109-780
77109-782
77685-205
2-Chloro-3,6-difluorobenzoic acid 98%
2-Chloro-3,6-difluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃ClF₂O₂ MW: 192.55 g/mol |
MDL Number:
MFCD01631441 CAS Number: 287172-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...